Learn More
Lenz Laborglasinstrumente™ Soda Lime Glass Beads
Click to view available options
Diameter (Metric) Outside:
2mm
3mm
4mm
5mm
6mm
Unit Size:
1kg
Specifications
Specifications
| Type | Beads |
| Material | Soda Lime Glass |
| Diameter (Metric) Outside | 5mm |
| Quantity | 1Pack |