Learn More
Brannan™ Soda Glass Range Finder Hydrometer
Product Code. 12831428
Change view
Click to view available options
Length (Metric):
320 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12831428 320 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12831428 Supplier Brannan™ Supplier No. 10/550/0
  • ONLINE EXCLUSIVE PRICE
    192.00 NOK
    177.04 NOK / Pack of 1
    Save 14.96 NOK
    7% Off
Estimated Shipment: 17-12-2026
Find the right hydrometer for your needs. Brannan™ Soda Glass Range Finder Hydrometers are frequently used to help select the appropriate hydrometer for subsequent accurate determinations.
Suitable for less accurate readings and are frequently used to help select the appropriate hydrometer for subsequent accurate determinations.
Specifications
Accuracy ±0.02g/mL
Increments 0.01 g/mL
Length (Metric) 320 mm
Specific Gravity Range 0.70 to 2.00 g/mL
Temperature of Standardization 15.6°C