Learn More
Jaytec Glass™ Soda Glass Milk and Cream Pipettes
Product Code. 12373448
Change view
Click to view available options
Volume (Metric):
0.5, 1 mL
1 mL
10 mL
10.94 mL
5 mL
Tolerance:
±0.02 mL
±0.03 mL
±0.05 mL
±0.08 mL
±0.15 mL
Type:
Amyl Alcohol Pipette
Milk Pipette
One-mark Blowout Pipette
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Tolerance Type Unit Size
12373448 1 mL ±0.05 mL Amyl Alcohol Pipette Each
12303448 0.5, 1 mL ±0.02 mL One-mark Blowout Pipette Each
12323448 5 mL ±0.08 mL One-mark Blowout Pipette Each
12333448 10 mL ±0.15 mL One-mark Blowout Pipette Each
12363448 10.94 mL ±0.03 mL Milk Pipette Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12373448 Supplier Jaytec Glass™ Supplier No. MP.687
  • ONLINE EXCLUSIVE PRICE
    106.00 NOK
    104.62 NOK / Pack of 1
    Save 1.38 NOK
    1% Off
Estimated Shipment: 03-02-2027
Soda Glass Milk and Cream Pipettes are for use with thick liquids.
  • Blowout pipettes with wide bore jets for thick liquids
  • The last drop in the jet must be blown-out except where shown
  • MP.688 (BS.696) NOT to be blown out
Specifications
Material Soda Lime Glass
Sterility Non-sterile
For Use With (Application) Used to deliver a precise volume of Amyl Alcohol in dairy testing
Graduations 1
Tolerance ±0.05 mL
Color Clear
Volume (Metric) 1 mL
Length (Metric) 250 mm
Type Amyl Alcohol Pipette
Disposable Reusable
For Use With (Equipment) Butyrometer