Learn More
Jaytec Glass™ Soda Glass Milk and Cream Pipettes
Description
- Blowout pipettes with wide bore jets for thick liquids
- The last drop in the jet must be blown-out except where shown
- MP.688 (BS.696) NOT to be blown out
Specifications
Specifications
| Material | Soda Lime Glass |
| Sterility | Non-sterile |
| For Use With (Application) | Delivery of a precise volume in milk/cream testing. Not to be blown out. |
| Graduations | 1 |
| Tolerance | ±0.03 mL |
| Color | Clear |
| Volume (Metric) | 10.94 mL |
| Length (Metric) | 335 mm |
| Type | Milk Pipette |
| Disposable | Reusable |
| Show More |