Learn More
Jaytec Glass™ Soda Glass Milk and Cream Pipettes
Click to view available options
Volume (Metric):
0.5, 1 mL
1 mL
10 mL
10.94 mL
5 mL
Tolerance:
±0.02 mL
±0.03 mL
±0.05 mL
±0.08 mL
±0.15 mL
Type:
Amyl Alcohol Pipette
Milk Pipette
One-mark Blowout Pipette
Unit Size:
Each
Description
- Blowout pipettes with wide bore jets for thick liquids
- The last drop in the jet must be blown-out except where shown
- MP.688 (BS.696) NOT to be blown out
Specifications
Specifications
| Material | Soda Lime Glass |
| Sterility | Non-sterile |
| For Use With (Application) | Delivery of a precise volume in milk/cream testing |
| Graduations | 1 |
| Tolerance | ±0.15 mL |
| Color | Clear |
| Volume (Metric) | 10 mL |
| Length (Metric) | 330 mm |
| Type | One-mark Blowout Pipette |
| Disposable | Reusable |
| Show More |