Learn More
Soda Glass Capillary Tube
Product Code. p-8008563
Click to view available options
Length (Metric) Tube:
75 mm
100 mm
Product Type:
Capillary Tube
Capillary Tube, Both Ends Sealed
Unit Size:
Pack of 100
Product Code. 12807685
Brand: S Murray SAMCO G119/02
Use these tubes for melting point determinations. Soda Glass Capillary Tubes provide excellent sample retention during centrifugation, and fill rapidly without affecting specimen integrity.
  • Soda glass capillary tubes
  • Outside diameter 1.8/2.0mm
  • Wall thickness 0.28/0.32mm
  • Packed in tubular cartons of 100
Specifications
Product Type Capillary Tube
Length (Metric) Tube 100 mm