Learn More
Soda Glass Capillary Tube
Product Code. 12897675
Change view
Click to view available options
Length (Metric) Tube:
75 mm
100 mm
Product Type:
Capillary Tube
Capillary Tube, Both Ends Sealed
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Tube Product Type Unit Size
12897675 100 mm Capillary Tube, Both Ends Sealed Pack of 100
12807685 100 mm Capillary Tube Pack of 100
12877675 75 mm Capillary Tube Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12897675 Supplier S Murray SAMCO Supplier No. G119/32
  • ONLINE EXCLUSIVE PRICE
    103.00 NOK
    77.31 NOK / Pack of 100
    Save 25.69 NOK
    24% Off
View all promotions
Estimated Shipment: 04-01-2027
Use these tubes for melting point determinations. Soda Glass Capillary Tubes provide excellent sample retention during centrifugation, and fill rapidly without affecting specimen integrity.
  • Soda glass capillary tubes
  • Outside diameter 1.8/2.0mm
  • Wall thickness 0.28/0.32mm
  • Packed in tubular cartons of 100
Specifications
Product Type Capillary Tube, Both Ends Sealed
Length (Metric) Tube 100 mm