missing translation for 'onlineSavingsMsg'
Learn More

SNUPN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15358-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16894504

  • 1611.84 NOK / 20µL
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein of unknown function. However, similarity to proteins in yeast and other species suggests that this protein may be an N-acetyltransferase.
Specifications

Specifications

SNUPN
Polyclonal
Unconjugated
SNUPN
KPNBL, RNA U transporter 1, RNUT1, SNUPN, snurportin 1, Snurportin1, SPN1
Rabbit
Antigen Affinity Chromatography
RUO
10073, 316108, 66069
-20°C
Liquid
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95149, Q68FP5, Q80W37
Snupn
SNUPN Fusion Protein Ag7604
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers