missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRPD2 Mouse anti-Human, Clone: 1F12B2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66111-1-IG-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the small nuclear ribonucleoprotein core protein family. It is required for pre-mRNA splicing and small nuclear ribonucleoprotein biogenesis. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| SNRPD2 | |
| Monoclonal | |
| 1.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P62316 | |
| SNRPD2 | |
| SNRPD2 Fusion Protein Ag7115 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 1F12B2 | |
| Unconjugated | |
| SNRPD2 | |
| Sm D2, SMD2, snRNP core protein D2, SNRPD2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 6633 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction