missing translation for 'onlineSavingsMsg'
Learn More

SNRPB2 Mouse anti-Human, Clone: 1D11E9, Proteintech

Mouse Monoclonal Antibody

1710.33 NOK - 4836.10 NOK

Specifications

Antigen SNRPB2
Clone 1D11E9
Concentration 1000 μg/mL
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17639786
View Documents
Proteintech
68095-1-IG-20UL
20 μL
1710.33 NOK
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17629786
View Documents
Proteintech
68095-1-IG-150UL
150 μL
4836.10 NOK
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SNRPB2 Monoclonal antibody apecifically detetects SNRPB2 antigen in Human Samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

SNRPB2
1000 μg/mL
Monoclonal
Liquid
Human
BC036737
SNRPB2
IgG1
Protein G purification
1D11E9
Western Blot, ELISA
Unconjugated
Mouse
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
6629
SNRPB2 fusion protein Ag5540
Primary
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.