missing translation for 'onlineSavingsMsg'
Learn More

SNIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14950-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16893874

  • 4593.76 NOK / 150µL
Estimated Shipment: 13-08-2024
to see stock.

Description

Description

Smad-binding peptide aptamers can be developed to selectively inhibit TGF-beta-induced gene expression.
Specifications

Specifications

SNIP1
Polyclonal
Unconjugated
SNIP1
RP3 423B22.3, SNIP1
Rabbit
Antigen Affinity Chromatography
RUO
313588, 76793, 79753
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5M9G6, Q8BIZ6, Q8TAD8
Snip1
SNIP1 Fusion Protein Ag6772
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers