Learn More
SNAPIN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10055-1-AP-150UL
 View more versions of this product
Product Code. 16836203
  • 4836.10 NOK / 150µL
Description
This antibody can recognize the 15-18 kDa (Monomer) and 30-36 kDa (Dimer) forms of SNAPIN.
The protein encoded by this gene is a coiled-coil-forming protein that associates with the SNARE (soluble N-ethylmaleimide-sensitive fusion protein attachment protein receptor) complex of proteins and the BLOC-1 (biogenesis of lysosome-related organelles) complex. Biochemical studies have identified additional binding partners. As part of the SNARE complex, it is required for vesicle docking and fusion and regulates neurotransmitter release. The BLOC-1 complex is required for the biogenesis of specialized organelles such as melanosomes and platelet dense granules. Mutations in gene products that form the BLOC-1 complex have been identified in mouse strains that are models of Hermansky-Pudlak syndrome. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
SNAPIN
Polyclonal
Unconjugated
SNAPIN
SNAP associated protein, SNAP25BP, SNAPAP, SNAPIN
Rabbit
Antigen Affinity Chromatography
RUO
20615, 23557, 295217
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.35 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95295, P60192, Q9Z266
Snapin
SNAPIN Fusion Protein Ag0101
150 μL
Primary
Human, Mouse, Rat