missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNAP25 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11558571
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11558571 100 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11558571 Supplier Invitrogen™ Supplier No. PA1740

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-740 detects synaptosomal associated protein 25 (SNAP-25) from bovine, mouse, rat and zebra fish (Danio rerio) samples. PA1-740 has been successfully used in Western blot and immunofluorescence procedures. By Western blot, this antibody detects a 25 kDa protein representing SNAP-25 in whole rat brain, mouse brain and zebra fish extracts. Immunofluorescence staining of SNAP-25 in cultured rat hippocampal cells with PA1-740 yields a pattern consistent with cytoplasmic and plasma membrane staining. PA1-740 immunizing peptide corresponds to the C-terminal amino acid residues 195-206 of mouse SNAP-25. This sequence is completely conserved between mouse, rat, human, chicken, and goldfish. PA1-740 immunizing peptide (Cat. # PEP-032) is available for use in neutralization and control experiments.

SNAP25 is a t-SNARE involved in the molecular regulation of neurotransmitter release. It may play a role in the synaptic function of specific neuronal systems. SNAP25 associates with proteins involved in vesicle docking and membrane fusion. It also regulates plasma membrane recycling through its interaction with CENPF. Mutations in the gene can results in myasthenic syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNAP25
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Snap25
Gene Accession No. P60879, P60881, Q17QQ3, Q5TZ66
Gene Alias ALMS1; bA416N4.2; Bdr; blind drunk; CG17676; CG17884; CG40452; CMS18; dJ1068F16.2; Dmel CG40452; Dmel_CG40452; etID17571.20; etID69167.20; fj42b02; fj45g04; fj48c02; FLJ23079; GENA70; HGNC:11132; resistance to inhibitors of cholinesterase 4 homolog; RIC4; RIC-4; SEC9; si:dkeyp-8f4.6; Snap; SNAP 25; snap25; SNAP-25; snap25.1; SNAP-25.1; snap25a; SNAP-25A; SNAP-25B; snp25; sp; SUP; super protein; synaptosomal associated protein-25; synaptosomal-associated 25 kDa protein; Synaptosomal-associated protein 25; synaptosomal-associated protein 25-A; synaptosomal-associated protein, 25 kDa; synaptosomal-associated protein, 25a; synaptosomal-associated protein, 25kDa; synaptosome associated protein 25; synaptosome associated protein 25a; synaptosome associated protein 25kDa; synaptosome-associated protein 25.1; synaptosome-associated protein 25a; wu:fj42b02; wu:fj45g04; wu:fj48c02
Gene Symbols Snap25, snap25a
Host Species Rabbit
Immunogen Synthetic Peptide:(C)- A(195) N Q R A T K M L G S G(206).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20614, 25012, 30712, 540853
Target Species Mouse, Rat, Bovine, Zebrafish
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.