missing translation for 'onlineSavingsMsg'
Learn More

JG Finneran Associates™ Snap Top Caps™

Product Code. 10656611
Change view
Click to view available options
Material:
PTFE, Butyl Rubber
PTFE, Silicone
Unit Size:
Pack of 1000
Product Code. Material unitSize
10656611 PTFE, Butyl Rubber Pack of 1000
10558122 PTFE, Silicone Pack of 1000
2 options
This item is not returnable. View return policy

Product Code. 10656611

Brand: JG Finneran Associates™ 524011

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color Clear
Material PTFE, Butyl Rubber
Closure Size 11 mm
For Use With (Equipment) Snap Ring/Crimp Top 12 x 32 mm Vials with 11 mm Crimp Finish
Product Type Snap Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.