Description
In contrast to crimp top and screw cap vials, snap ring vials are chosen strictly for convenience. Snap caps may be adequate for applications that do not require heating of the sample above room temperature and where moderate sealing performance can be tolerated.
- Packed in zip-lock PE-bags with tear-off foil strip to ensure tamper-evidence and re-closeability
Specifications
Specifications
| Color (Seal) | Red/White/Red |
| Diameter (Metric) Inner Neck | 11 mm |
| Color | Transparent |
| Diameter (Metric) Outer | 12.09 mm |
| Material | Polypropylene, Polyethylene |
| Septum | PTFE/Silicone/PTFE |
| For Use With (Application) | High Performance Liquid Chromatography (HPLC) |
| Hardness | 45 ± 5 shore A |
| Quantity | 100 |
| Thickness (Metric) | 1.02 mm |
| Show More |