missing translation for 'onlineSavingsMsg'
Learn More

SNAI1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16844705 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16844705 20 μL 20µL
16834705 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16844705

Brand: Proteintech 261831AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Zinc finger protein SNAI1 (SNAIL)is a zinc finger transcriptional repressor which down regulates the expression of the ectodermal gene within the mesoderm and functions to form the mesoderm and neural crest. It is reported that SNAI1 acts as an important factor of tumor invasion as evidenced by its role in E-cadherin down-regulation and induction of epithelial-mesenchymal transition (EMT). In human breast cancer, the expression of SNAI1 and/or the homologous SNAI2 (Slug) has been associated with E-cadherin repression, local or distant metastasis, tumor recurrence, or poor prognosis in different tumor series. It is also reported that Snail 1 protein in the stromamay be anew putative prognosis marker for colon tumors.

Especificaciones

Antigen SNAI1
Applications Immunofluorescence, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SNAI1
Gene Accession No. O95863, Q02085
Gene Alias Protein sna, Protein snail homolog 1, SLUGH2, SNA, SNAH, SNAI1, snail homolog 1 (Drosophila), SNAIL1, Zinc finger protein SNAI1
Gene Symbols SNAI1
Host Species Rabbit
Immunogen SNAI1 Fusion Protein Ag24248
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20613, 6615
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.