Learn More
SMURF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55175-1-AP-20UL
 View more versions of this product
Product Code. 16856655
  • 1710.33 NOK / 20µL
Description
The antibody is specific to SMURF1.
Members of the transforming growth factor-beta (TGFB) family signal through type I and type II serine/threonine-kinase receptors, which in turn activate the SMAD signaling pathway. Bone morphogenetic protein (BMP) receptors target SMAD1, SMAD5, and SMAD8, whereas receptors for activin and TGFB (e. g., ACVR1 and TGFBR1, respectively) target SMAD2 and SMAD3. Phosphorylation of these receptor-regulated SMADs induces their association with the common-partner SMAD, SMAD4. Smurf1, a HECT domain E3 ubiquitin ligase, regulates cell polarity and protrusive activity and is required to maintain the transformed morphology and motility of a tumor cell. Atypical protein kinase C-zeta (PKC2), an effector of the Cdc42/Rac1-PAR6 polarity complex, recruits Smurf1 to cellular protrusions, where it controlled the local level of RhoA. Smurf1 thus links the polarity complex to degradation of RhoA in lamellipodia and filopodia to prevent RhoA signaling during dynamic membrane movements.
Specifications
Specifications
SMURF1
Polyclonal
Unconjugated
SMURF1
hSMURF1, KIAA1625, SMURF1
Rabbit
Antigen Affinity Chromatography
RUO
57154, 690516, 75788
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.9 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9CUN6, Q9HCE7
SMURF1
Peptide
20 μL
Primary
Mouse, Human, Rat