Learn More
Sartorius Smooth Technical Filter Papers, Discs
This Smooth Technical Filter Paper grade is generally used for analytical and technical purposes.
97.90 NOK - 494.00 NOK
Product Code | Brand | Diameter (Metric) | Grade | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Grade | Price | Quantity & Availability | ||||
15982910
|
Sartorius
FT-3-309-030 |
30 mm | 4 b |
97.90 NOK
Pack of 100 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
||||
16600583
|
Sartorius
FT-3-328-270 |
270 mm | 100/N |
494.00 NOK
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
Description
- Made of refined pulp and cotton linters with an >95% α-cellulose content
- Ash content between 0.1 to 0.15% (grade 100/N <0.1%)
- Wet-strengthened
- Supplied in rolls, sheets, discs and folded filters as well as customer-specific cuts
Specifications
Cotton Linters, Refined Pulp | |
Cellulose |