Learn More
Sartorius Smooth Technical Filter Papers
This Smooth Technical Filter Paper grade is generally used for analytical and technical purposes.
150.00 NOK - 3550.00 NOK
Specifications
Material | Cotton Linters, Refined Pulp |
---|---|
Material (Membrane) | Cellulose |
Type | Smooth Filter Paper |
Fit for Purpose | Certified Quality |
Product Code | Brand | Dimensions (L x W) | Grade | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Grade | Price | Quantity & Availability | ||||
15952720
|
Sartorius
FT-1-H079-040020 |
40 x 20 mm (1.6 x 0.8 in.) | H079 |
755.00 NOK
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
15902780
|
Sartorius
FT-2-356-089140 |
89 × 140 mm (3.5 x 5.5 in.) | C 140 |
150.00 NOK
Each |
In Stock
Log in to see stock. |
||||
15922780
|
Sartorius
FT-2-356-445572 |
445 × 572 mm (17.5 x 22.5 in.) | C 140 |
3550.00 NOK
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
15952760
|
Sartorius
FT-2-326-160210 |
160 x 210 mm (6.3 x 8.3 in.) | 69 K |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Made of refined pulp and cotton linters with a >95% α-cellulose content
- Ash content between 0.1 to 0.15%
- Wet-strengthened
Specifications
Cotton Linters, Refined Pulp | |
Cellulose | |
Smooth Filter Paper | |
Certified Quality |