missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMCR7L/MID51 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20164-1-AP-150UL
This item is not returnable.
View return policy
Description
The specific function of this protein remains unknown.Specifications
| SMCR7L/MID51 | |
| Polyclonal | |
| Unconjugated | |
| MIEF1 | |
| HSU79252, MID51, MIEF1, SMCR7L | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 239555, 315141, 54471 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5XIS8, Q8BGV8, Q9NQG6 | |
| Mief1 | |
| SMCR7L/MID51 Fusion Protein Ag13775 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction