Learn More
Techsit™ Smartsit High Laboratory Chair
Description
- Easy to clean
- Adjustable foot ring on glides
- Hard wearing
- Ergonomically designed and contoured seat and back
Specifications
Specifications
| Color | Black |
| Adjustable (Height) | Yes |
| Back Style | Ergonomic |
| Certifications/Compliance | Cleanroom Class 5 |
| Finish | Polyurethane |
| Frame Material | Steel |
| Seating Height | 590 to 830 mm |
| Warranty | 5 Year |
| Weight (English) | 15 kg |
| Wheel Type | Glide |
| Show More |