Learn More
Tork SmartOne™ Mini Toilet Roll White T9
Product Code. 17871398 Shop All Tork Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17871398 White Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17871398 Supplier Tork Supplier No. 472193
  • ONLINE EXCLUSIVE PRICE
    697.00 NOK
    649.50 NOK / Pack of 12
    Save 47.50 NOK
    6% Off
View all promotions
 Restricted Item
Estimated Shipment: 02-12-2026
The Tork SmartOne Mini Toilet Roll System uniquely delivers one hygienic sheet at a time, helping to reduce consumption by up to 40% compared to traditional jumbo roll dispensers, which means more visits per roll.
  • Perfectly balance costs and maintenance with our SmartOne™ toilet tissue.
  • Improve hygiene levels with single-sheet dispensing - guests touch only what they take.
  • Reduce the risk of paper shortages thanks to this high-capacity dispenser.
  • Save staff time with the SmartCore™ core removal for fast and easy refills.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Toilet Tissue Roll
Length (English) 122.05 yd.
Length (Metric) 111.6 m
Width (English) 5.28 in.
Width (Metric) 13.4 cm
Color White
Ply 2-ply