Learn More
Thermo Scientific™ SMART SPME Fibers for TriPlus™ RSH SMART Autosamplers
Product Code. p-7222042 Shop All Thermo Scientific Products
Click to view available options
Phase:
Acrylate, 85 μm
Assorted
Carbon WR/PDMS, 95 μm
DVB/Carbon WR/PDMS, 50/30 μm
DVB/PDMS, 65 μm
PDMS, 100 μm
PDMS, 30 μm
PDMS, 7 μm
Quantity:
1/Pk.
3/Pk.
5/Pk.
Unit Size:
Each
Pack of 3
Pack of 5
Product Code. 17797454
Brand: Thermo Scientific™ 36SP01P1SM
Only null left
Add to basket
Add to basket
Track part and lot numbers, usage range, history, and more for SMART SPME Fibers on the TriPlus™ RSH SMART Autosampler.
Track information such as part number, lot number, usage range, and history for Thermo Scientific™ SMART SPME Fibers installed on TriPlus™ RSH SMART Autosamplers. Each fiber is equipped with an ID chip, which stores this usage-based information for fast, solvent-free, and automated sample preparation. When installed on the TriPlus RSH SMART Autosampler, the information for SPME Fibers can be directly accessed and reported via Thermo Scientific™ Chromeleon™ CDS software.
Automate solvent-free solid phase micro extraction (SPME) sample preparation by performing sample extraction and enrichment in a single step with our SMART SPME Fibers.
SMART SPME Fibers also offer the additional benefit of tracking usage information. SMART SPME Fibers are compatible with all TriPlus™ RSH instruments as well as PAL platforms and AOC-6000 series instruments.
SMART Technology Benefits
  • Gain higher confidence in analytical results
  • Ensure error free usage of SPME Fibers through automatic identification
  • Support GLP accreditation with usage history traceability
  • Optimize consumption by monitoring of usage conditions and lifetime
  • Facilitate SPME Fibers management thanks to color-coded health notification dashboard integrated in Chromeleon™ CDS
TRUSTED_SUSTAINABILITY
Specifications
Thickness (Metric) 7 μm
Length (Metric) Phase Length: 10 mm
Quantity 1/Pk.
Description SMART SPME Fiber: PDMS (Polydimethylsiloxane), 7 μm, Green, 1/Pk.
Type Fiber
Phase PDMS, 7 μm
Color Green
Diameter (Metric) Outer 23 Gauge