Learn More
Thermo Scientific™ SMART SPME Arrows for TriPlus™ RSH SMART Autosamplers
Product Code. p-7222041 Shop All Thermo Scientific Products
Click to view available options
Phase:
Acrylate, 100 μm
Assorted
Carbon WR/PDMS, 120 μm
DVB/C-WR/PDMS, 120 μm
DVB/PDMS, 120 μm
PDMS, 100 μm
PDMS, 250 μm
Quantity:
Pk. of 1
Pk. of 3
Pk. of 5
Unit Size:
Each
Pack of 3
Pack of 5
Product Code. 17767444
Brand: Thermo Scientific™ 36SA10A1SM
Only null left
Add to basket
Add to basket
Track part and lot numbers, usage range, history, and more for Thermo Scientific™ SMART SPME Arrows on the TriPlus™ RSH SMART Autosampler.
Track information such as part number, lot number, usage range, and history for Thermo Scientific™ SMART SPME Arrows installed on TriPlus™ RSH SMART Autosamplers. Each arrow is equipped with an ID chip, which stores this usage-based information for fast, solvent-free, and automated sample preparation. When installed on the TriPlus RSH SMART Autosampler, the information for SMART SPME Arrows can be directly accessed and reported via Thermo Scientific™ Chromeleon™ CDS software.
Automate solvent-free solid phase micro extraction (SPME) sample preparation by performing sample extraction and enrichment in a single step with our SMART SPME Arrows.
SMART SPME Arrows also offer the additional benefit of tracking usage information. SMART SPME Arrows are compatible with all TriPlus™ RSH instruments as well as PAL platforms and AOC-6000 series instruments.
SMART Technology Benefits
  • Gain higher confidence in analytical results
  • Ensure error free usage of SPME Arrows through automatic identification
  • Support GLP accreditation with usage history traceability
  • Optimize consumption by monitoring of usage conditions and lifetime
  • Facilitate SPME Arrow management thanks to color-coded health notification dashboard integrated in Chromeleon™ CDS
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) Phase Length: 20 mm
Quantity Pk. of 1
Description SMART SPME Arrow 1.1 mm: Acrylate (Polyacrylate), 100 μm, Grey, 1/Pk.
Type Arrow
Phase Acrylate, 100 μm
Color Gray
Diameter (Metric) Outer 1.1 mm