Learn More
Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Streptavidin
Product Code. p-9078061 Shop All Thermo Scientific Products
Click to view available options
Description:
Magnetic, SOLAμ Collection Plate, with Soluble Trypsin
Magnetic, with Soluble Trypsin
Magnetic, without Soluble Trypsin
Unit Size:
1 set
Product Code. 30130375
Brand: Thermo Scientific™ 60118101
Perform simple and easy streptavidin immunocapture for protein sample cleanup prior to protein digestion or analysis.
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Streptavidin. These kits were designed to remove the challenges often associated with the process of protein cleanup by providing a simple and easy capture workflow.
The kit contains beads with an attached streptavidin moiety enabling a strong streptavidin-biotin bond with the appropriate biotinylated molecule with specific affinity for the target biomolecule. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once completed subsequent digestion can be facilitated with SMART Digest enzymes if required.
TRUSTED_SUSTAINABILITY
Specifications
Description Magnetic, without Soluble Trypsin
Capture Streptavidin
Cleavage C-terminal side of lysine and arginine amino acids
Enzyme Trypsin
Format Bulk Magnetic
For Use With (Application) Immunocapture Sample Preparation
Product Line SMART Digest
Type Immunocapture
Includes Bulk Magnetic SMART Digest Resin