missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCD1 Mouse anti-Human, Clone: 2F2G6, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | SMARCD1 |
|---|---|
| Clone | 2F2G6 |
| Concentration | 1.05 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16897695
|
Proteintech
66116-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16887695
|
Proteintech
66116-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and has sequence similarity to the yeast Swp73 protein. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| SMARCD1 | |
| 1.05 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| BAF60A, BRG1 associated factor 60A, CRACD1, Rsc6p, SMARCD1, SWI/SNF complex 60 kDa subunit | |
| SMARCD1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 2F2G6 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q96GM5 | |
| 6602 | |
| SMARCD1 Fusion Protein Ag19100 | |
| Primary | |
| -20°C | |
| SMARCD1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title