missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17722-1-AP-150UL
This item is not returnable.
View return policy
Description
The transcription factor ATF-2 (also called CRE-BP1) binds to both AP-1 and CRE DNA response elements and is a member of the ATF/CREB family of leucine zipper proteins. ATF-2 interacts with a variety of viral oncoproteins and cellular tumor suppressors and is a target of the SAPK/JNK and p38 MAP kinase signaling pathways. Various forms of cellular stress, including genotoxic agents, inflammatory cytokines and UV irradiation, stimulate the transcriptional activity of ATF-2. Cellular stress activates ATF-2 by phosphorylation of Thr69 and Thr71. Both SAPK and p38 MAPK have been shown to phosphorylate ATF-2 at these sites in vitro and in cells transfected with ATF-2.Specifications
| SMARCC1 | |
| Polyclonal | |
| Unconjugated | |
| SMARCC1 | |
| BAF155, BRG1 associated factor 155, CRACC1, Rsc8, SMARCC1, SRG3, SWI3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20588, 301020, 6599 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P97496, Q92922 | |
| SMARCC1 | |
| SMARCC1 Fusion Protein Ag12109 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction