missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12799-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is a rabbit polyclonal antibody raised against residues near the N terminus of human SMAP1.
The protein encoded by this gene is similar to the mouse stromal membrane-associated protein-1. This similarity suggests that this human gene product is also a type II membrane glycoprotein involved in the erythropoietic stimulatory activity of stromal cells. Alternate splicing results in multiple transcript variants encoding different isoforms.Specifications
| SMAP1 | |
| Polyclonal | |
| Unconjugated | |
| SMAP1 | |
| small ArfGAP 1, SMAP 1, SMAP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 60682, 684800, 98366 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.48 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8IYB5, Q91VZ6 | |
| SMAP1 | |
| SMAP1 Fusion Protein Ag3544 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction