Learn More
Sampling Systems™ Small Volume Medicine Cups
Product Code. 15809721 Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
30 mL
Unit Size:
Each
Product Code. 15809721
Brand: Sampling Systems™ A76430
Only null left
Add to basket
Add to basket
Designed with a flat base to make them ideal for weigh scales. Sampling Systems™ Small Volume Medicine Cups are ideal for holding small amounts of tablets, granules or powders.
  • 304 Stainless Steel
  • Crevice free
  • Polished
Specifications
Diameter (Metric)Top 50mm
Capacity (Metric) 30 mL
Material Stainless Steel