Learn More
Tork Small Pack Dispenser W8
Product Code. 30243867 Shop All Tork Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30243867 White Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30243867 Supplier Tork Supplier No. 655100
  • ONLINE EXCLUSIVE PRICE
    516.53 NOK
    461.09 NOK / Pack of 4
    Save 55.44 NOK
    10% Off
Estimated Shipment: 21-12-2026
The Tork Small Pack dispensers for folded wipes in Performance design provide space-saving convenience, hygienic protection and one-at-a time dispensing to reduce consumption and waste.
  • Organise tight workplaces and save space with this wall-mounted dispenser.
  • Avoid wasting time as folded wipes are within easy reach.
  • Provide hygienic protection as staff will only touch the wipes they use.
  • Reduce waste and save money, thanks to one-at-a-time dispensing.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Wipe Holder
Color White
Alternate Part Number 7322540862447
Length (Metric) 21.4 cm
Width (Metric) 10.7 cm
Height (Metric) 14.6 cm
Holds Wipe Containers
Material Plastic
Mounting Wall Mount
No. of Compartments 1
Product Style Open Top
Model Performance