Learn More
Hamilton™ Small Hub Removable Needles
Product Code. 12702257
Click to view available options
Needle Gauge:
18 G
22 G
22s G
23s G
26 G
26s G
28 G
29 G
30 G
31 G
32 G
33 G
Needle Point Style:
2, 3, or 4
5 or AS
Unit Size:
Each
Pack of 6
Product Code. 12702257
Brand: Hamilton™ 780306
Available in custom lengths and point styles to fit your laboratory needs. Hamilton™ Small Hub Removable Needles are manufactured from stainless steel.
  • Autoclavable
  • Standard needle length is 2"/51mm
  • Can be used with (RN)-style syringes that are 100μL and smaller
Specifications
Material Stainless Steel
Needle Gauge 29 G
Diameter (Metric) Outer 0.34 mm
Product Type Removable Custom Needle
Length (Metric) Needle 9.52 to 304 mm
Needle Point Style 2, 3, or 4
For Use With (Equipment) Small Removable Syringes (100μL and smaller)