Learn More
Hamilton™ Small Hub Removable Needles
Product Code. 10492005
Change view
Click to view available options
Needle Gauge:
18 G
22 G
22s G
23s G
26 G
26s G
28 G
29 G
30 G
31 G
32 G
33 G
Needle Point Style:
2, 3, or 4
5 or AS
Unit Size:
Each
Pack of 6
Product Code. Needle Gauge Needle Point Style unitSize
10492005 26s G 2, 3, or 4 Pack of 6
10595855 26s G 5 or AS Pack of 6
11530332 33 G 2, 3, or 4 Pack of 6
10798204 22s G 2, 3, or 4 Pack of 6
11573294 22 G 5 or AS Pack of 6
12386439 18 G 2, 3, or 4 Pack of 6
11510332 31 G 2, 3, or 4 Pack of 6
10698205 26 G 2, 3, or 4 Pack of 6
15927335 26 G 2, 3, or 4 Each
12316299 23s G 5 or AS Pack of 6
11500332 28 G 2, 3, or 4 Pack of 6
10725873 22s G 5 or AS Pack of 6
11532525 22 G 2, 3, or 4 Pack of 6
12702257 29 G 2, 3, or 4 Pack of 6
10776063 30 G 2, 3, or 4 Pack of 6
11520332 32 G 2, 3, or 4 Pack of 6
10140888 26 G 5 or AS Pack of 6
17 options
Product Code. 10492005
Brand: Hamilton™ 780404
Available in custom lengths and point styles to fit your laboratory needs. Hamilton™ Small Hub Removable Needles are manufactured from stainless steel.
  • Autoclavable
  • Standard needle length is 2"/51mm
  • Can be used with (RN)-style syringes that are 100μL and smaller
Specifications
Material Stainless Steel
Needle Gauge 26s G
Diameter (Metric) Inner 0.13 mm
Diameter (Metric) Outer 0.47 mm
Product Type Removable Custom Needle
Needle Point Style 2, 3, or 4
For Use With (Equipment) Small Removable Syringes (100μL and smaller)