Description
The SMAD7 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human SMAD7. This antibody recognizes human, mouse, rat antigen. The SMAD7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
| Antigen | SMAD7 |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Concentration | 540 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Quantity | 20 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 4092 |
| Target Species | Human, Rat, Mouse |
| Show More |