Learn More
Tocris Bioscience™ SM 102
Product Code. 30537733
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30537733 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 30537733 Supplier Tocris Bioscience™ Supplier No. 8909/25
Stock Up and Save This Summer
Use promo code 27950
View all promotions
 Restricted Item
Estimated Shipment: 23-09-2026
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Cationic ionizable lipid
SM 102 is an ionizable cationic lipid (pKa = 6.68) used in the formulation of lipid nanoparticles (LPNs) for the delivery of mRNA in vitro and in vivo. Administration of LNPs containing SM 102 induces hepatic luciferase expression in mice. SM 102 is commonly formulated with cholesterol (Cat. No. 7945), DSPC (Cat. No. 7943), and DMG-PEG 2000 (Cat. No. 7944).
For more information on LNPs and available components see our Lipid Nanoparticles page .
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°C
Quantity 25 mg