Learn More
Karl Hecht™ Assistent™ Sloping Stem Glass Funnels
Product Code. 10398971
Change view
Click to view available options
Diameter (Metric)Top:
80mm
100mm
150mm
200mm
450mm
550mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top unitSize
10398971 100mm Each
13108365 200mm Each
10479241 450mm Each
10247391 550mm Each
10093811 80mm Each
10083761 150mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10398971 Supplier Karl Hecht™ Supplier No. 44000040
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Karl Hecht™ Assistent™ Sloping Stem Glass Funnels are manufactured from clear soda-lime glass.
  • Slope angle of 60°
  • Available in a range of diameters
Specifications
Diameter (Metric)Top 100mm
Material Glass
Angle 60°
For Use With (Application) Laboratory Applications