Learn More
Thermo Scientific™ SLIPFREE Ferrules
Product Code. 10404783 Shop All Thermo Scientific Products
Change view
Click to view available options
Material:
PEEK
Vespel
Unit Size:
Each
Product Code. Material unitSize
10404783 PEEK Each
10140643 Vespel Each
2 options
Product Code. 10404783
Brand: Thermo Scientific™ 36023
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ SLIPFREE Ferrules are for use with SLIPFREE connectors for HPLC columns.
Thermo Scientific™ SLIPFREE Ferrules are for use with SLIPFREE connectors for HPLC columns.
  • Vespel ferrules replace the standard Vespel ferrules supplied with SLIPFREE column connectors
  • Kel-F and PEEK are offered for applications in which Vespel is not suitable
TRUSTED_SUSTAINABILITY
Specifications
Type Sealing Ferrule
Description PEEK sealing ferrule
For Use With (Equipment) SLIPFREE Connectors for HPLC Columns
Material PEEK