Learn More
3M™ Safety-Walk™ Slip Resistant General Purpose Tape, Series 600
Product Code. p-7213721
Change view
Click to view available options
Color:
Black
Transparent
Yellow
Width (Metric):
19 mm
25 mm
51 mm
102 mm
152 mm
305 mm
508 mm
914 mm
Unit Size:
1 roll
2 rolls
4 rolls
Each
Pack of 2
Pack of 4
Product Code. Color Width (Metric) unitSize
17108690 Black 19 mm 4 rolls
17118780 Transparent 19 mm 4 rolls
17148760 Transparent 152 mm 1 roll
17128680 Black 305 mm Each
17158760 Transparent 305 mm 1 roll
17138780 Yellow 305 mm 1 roll
17128790 Black 508 mm 1 roll
17158680 Black 914 mm Each
17128780 Yellow 19 mm 4 rolls
17158700 Black 25 mm Pack of 4
17168590 Transparent 25 mm 4 rolls
17188600 Yellow 25 mm 4 rolls
17118560 Transparent 51 mm Pack of 2
17198670 Yellow 51 mm 2 rolls
17108680 Transparent 102 mm 1 roll
17188680 Black 152 mm 1 roll
16 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17108690
Marca: 3M™ Safety-Walk™ 7100032939
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Mineral coated, slip resistant material features a highly durable surface. Low profile design helps reduce tripping hazards.
  • Anti-slip tape for light to heavy shoe-traffic areas
  • High quality abrasive particles to ensure long-lasting friction in any application
  • Highly durable surface withstands the rigors of equipment traffic
  • High performance, pressure-sensitive adhesive for easy application
  • Custom shapes and sizes are also available
  • NFSI High Traction Certification
TRUSTED_SUSTAINABILITY
Specifica
Dimensions (L x W) 18.3 m x 19 mm
Finish Mineral-coated
For Use With (Application) Light to Heavy Traffic Areas
Length (English) 60 ft.
Length (Metric) 18.3 m
Packaging Quantity 4 Pk.
Width (English) 0.75 in.
Width (Metric) 19 mm
Adhesive Type Pressure Sensitive Adhesive
Color Black
Type Floor Tape
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.