Learn More
3M™ Safety-Walk™ Slip Resistant Coarse Tape, Series 700
Description
- Anti-slip tape for extreme and heavy traffic areas
- Extra-large abrasive particles resist clogging in muddy or oily conditions
- Can be painted without losing friction properties
- High performance adhesive
- Suitable for use on agriculture and forestry equipment, road building, civil engineering, lorries, tractors, construction vehicles, cranes, and building equipment
Specifications
Specifications
| Certifications/Compliance | DIN 51130: R 13, EN 13501 1: Bfl s1 |
| Description | Extra Strong (Type 4) |
| Dimensions (L x W) | 18.3 m x 51 mm |
| For Use With (Application) | Additional Slip Protection in Extreme, Heavily Frequented Areas |
| Length (Metric) | 18.3 m |
| Packaging Quantity | 1 Roll |
| Width (Metric) | 51 mm |
| Adhesive Type | Pressure Sensitive Adhesive |
| Color | Black |