Learn More
Syngene™ Slimline Flat White Light Panel - Fold Down
Product Code. 13191514
Change view
Click to view available options
Quantity:
1
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13191514 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13191514 Supplier Syngene™ Supplier No. WLP2014GX
  • ONLINE EXCLUSIVE PRICE
    7325.00 NOK
    6880.92 NOK / Pack of 1
    Save 444.08 NOK
    6% Off
Estimated Shipment: 26-01-2027
Syngene™ Slimline Flat White Light Panel - Fold Down is for use with G:BOX F3 and G:BOX Chemi XRQ systems
  •  Size: 20 x 14cm
  • Fold down white light pad with brackets
  • Includes a fold down tray, built into a standard size G:BOX darkroom cabinet.
Specifications
For Use With (Equipment) Syngene Gbox F3 and XRQ
Quantity 1