Learn More
Thermo Scientific™ Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Product Code. 11399311 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
Boxes (2 in.)
Boxes (3 in.)
Matrix 2D Internally Threaded Tubes (200 μL)
Matrix 2D Tubes (1 mL)
Matrix 2D Tubes (500 μL)
Microplates
Microplates with Lids
Nunc or Matrix 2D Tubes (5 mL)
Tubes
Positions:
12 Boxes/Rack
16 Boxes/Rack
16 Plates/Rack
18 Boxes/Rack
21 Boxes/Rack
24 Boxes/Rack
24 Plates/Rack
28 Boxes/Rack
28 Plates/Rack
30 Plates/Rack
36 Boxes/Rack
42 Boxes/Rack
44 Plates/Rack
48/Box
66 Plates/Rack
70 Plates/Rack
84 Plates/Rack
9 Boxes/Rack
96/Box
For Use With (Equipment):
EXF240, HFU240B (13 cu.ft.)
EXF240, HFU240B (5 shelf) (13 cu.ft.)
EXF320, EXF400, HFU320B, HFU400B (17.3, 23 cu.ft.)
EXF320, EXF400, HFU320B, HFU400B (5 shelf) (17.3, 23 cu.ft.)
EXF600, HFU600B (28 cu.ft.)
Revco UxF, HERAFreeze HFU-T (14.9, 19.4, 24.1, 28.8, 33.5 cu.ft.)
Upright Freezers (13 cu.ft.)
Upright Freezers (17.3, 23 cu.ft.)
Upright Freezers (17.3, 23, 28.8 cu.ft.)
Unit Size:
Each
Codice prodotto. Holds Positions For Use With (Equipment) unitSize
11399311 Boxes (2 in.) 12 Boxes/Rack Upright Freezers (13 cu.ft.) Each
15140509 Tubes 96/Box EXF240, HFU240B (13 cu.ft.) Each
15150509 Matrix 2D Internally Threaded Tubes (200 μL) 36 Boxes/Rack EXF320, EXF400, HFU320B, HFU400B (17.3, 23 cu.ft.) Each
15130509 Tubes 96/Box EXF600, HFU600B (28 cu.ft.) Each
15111045 Matrix 2D Internally Threaded Tubes (200 μL) 42 Boxes/Rack Revco UxF, HERAFreeze HFU-T (14.9, 19.4, 24.1, 28.8, 33.5 cu.ft.) Each
11359291 Microplates 16 Plates/Rack EXF240, HFU240B (5 shelf) (13 cu.ft.) Each
11711101 Microplates 24 Plates/Rack EXF320, EXF400, HFU320B, HFU400B (5 shelf) (17.3, 23 cu.ft.) Each
15140539 Matrix 2D Tubes (500 μL) 28 Boxes/Rack EXF600, HFU600B (28 cu.ft.) Each
11379291 Microplates 28 Plates/Rack Revco UxF, HERAFreeze HFU-T (14.9, 19.4, 24.1, 28.8, 33.5 cu.ft.) Each
15160509 Tubes 96/Box EXF240, HFU240B (13 cu.ft.) Each
15170509 Matrix 2D Tubes (1 mL) 18 Boxes/Rack EXF320, EXF400, HFU320B, HFU400B (17.3, 23 cu.ft.) Each
15180509 Matrix 2D Tubes (1 mL) 21 Boxes/Rack EXF600, HFU600B (28 cu.ft.) Each
15190509 Matrix 2D Tubes (1 mL) 21 Boxes/Rack Revco UxF, HERAFreeze HFU-T (14.9, 19.4, 24.1, 28.8, 33.5 cu.ft.) Each
15100519 Tubes 48/Box EXF240, HFU240B (13 cu.ft.) Each
15110519 Nunc or Matrix 2D Tubes (5 mL) 24 Boxes/Rack EXF320, EXF400, HFU320B, HFU400B (17.3, 23 cu.ft.) Each
15130519 Tubes 48/Box EXF600, HFU600B (28 cu.ft.) Each
15120519 Nunc or Matrix 2D Tubes (5 mL) 28 Boxes/Rack Revco UxF, HERAFreeze HFU-T (14.9, 19.4, 24.1, 28.8, 33.5 cu.ft.) Each
11369311 Microplates with Lids 70 Plates/Rack Upright Freezers (13 cu.ft.) Each
11389311 Microplates with Lids 84 Plates/Rack Upright Freezers (17.3, 23 cu.ft.) Each
11309321 Boxes (3 in.) 9 Boxes/Rack Upright Freezers (13 cu.ft.) Each
11319321 Boxes (2 in.) 16 Boxes/Rack Upright Freezers (17.3, 23 cu.ft.) Each
11359321 Boxes (3 in.) 12 Boxes/Rack Upright Freezers (17.3, 23 cu.ft.) Each
11369321 Boxes (2 in.) 16 Boxes/Rack Upright Freezers (17.3, 23 cu.ft.) Each
11379321 Boxes (2 in.) 12 Boxes/Rack Upright Freezers (13 cu.ft.) Each
11389321 Microplates 66 Plates/Rack Upright Freezers (17.3, 23, 28.8 cu.ft.) Each
11399321 Microplates 44 Plates/Rack Upright Freezers (13 cu.ft.) Each
10731310 Boxes (3 in.) 9 Boxes/Rack Upright Freezers (13 cu.ft.) Each
11376311 Microplates 30 Plates/Rack Upright Freezers (17.3, 23, 28.8 cu.ft.) Each
11329321 Boxes (2 in.) 12 Boxes/Rack Upright Freezers (17.3, 23 cu.ft.) Each
29 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11399311
Marca: Thermo Scientific™ 398324
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Maximize tube storage capacity per freezer with a racking solution for five inner-door, ultra-low temperature freezers.
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for TSX/TDE 5 shelf Series Freezers as well as well as previous generation Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • Warranty varies by country, please contact us for details
TRUSTED_SUSTAINABILITY
Specifica
Description 2 in. Box Racks - Adjustable side access rack
Color Silver
Holds Boxes (2 in.)
Material Stainless Steel
Positions 12 Boxes/Rack
For Use With (Equipment) Upright Freezers (13 cu.ft.)
Width (English) 5.4 in.
Height (English) 9.4 in.
Depth (English) 16.5 in.
Depth (Metric) 41.9 cm
Product Type Side Access Freezer Rack (Adjustable)
Shelves 4 Tiers
Height (Metric) 23.9 cm
Width (Metric) 13.7 cm
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.