Learn More
Fisherbrand™ Sliding Device
Product Code. 11552703
Click to view available options
For Use With (Equipment):
Inverted Biological Microscope FB69196
Unit Size:
Each
Product Code. 11552703
Brand: Fisherbrand™ 3699.5406
Use this accessory with phase contrast for inverted biological microscopes. Fisherbrand™ Sliding Devices are ergonomically designed for everyday use.
  • Sliding Device
  • Use with phase contrast
  • For inverted biological microscope FB69196
Specifications
Type Sliding Device for Phase Contrast
For Use With (Equipment) Inverted Biological Microscope FB69196