Learn More
Hecht Karl™ Slides
Description
- Microscope slides/plates with one or more cavities.
- Ground edges.
Specifications
Specifications
| Length (Metric) | 76 mm |
| Width (Metric) | 26 mm |
| Type | Slide |
| Well Count | 3 |
| Quantity | 1 Pack |
| Dimensions (L x W) | 76 x 26 mm |