Learn More
Epredia™ SlideMate™ Accessories
Product Code. 12618346 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Digital Imager
Keyboard Interface
LabWriter Software
Mini PC Package, clamp arm
Mini PC Package, stand
Mini PC, imager, stand
Thermal Transfer Ribbon
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
12618346 Thermal Transfer Ribbon Each
12933815 Mini PC Package, clamp arm Each
12943815 Mini PC Package, stand Each
12666516 LabWriter Software Each
12913805 Digital Imager Each
12903825 Keyboard Interface Each
12923815 Mini PC, imager, stand Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12618346 Supplier Epredia™ Supplier No. B81320041
  • ONLINE EXCLUSIVE PRICE
    2785.00 NOK
    2168.36 NOK / Pack of 1
    Save 616.64 NOK
    22% Off
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
Enhance and optimize the Epredia™ SlideMate™ Slide Printer with Epredia™ SlideMate Slide Printer Accessories.
Accessories include:
  • Collection system Collection trays (box of 4)
  • Hopper
  • Hot foil tape, box of 5 (roll of tape prints approx. 20,000 cassettes)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Thermal Transfer Ribbon
For Use With (Equipment) SlideMate™ Slide Printer
Quantity Box of 1 (prints approx. 12,000 slides per roll of tape)