Learn More
Epredia™ Slide Mailer
Product Code. 10402614 Shop All Epredia Products
Product Code. 10402614
Brand: Epredia™ OV20031
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Five slide-box for microscope slides
  • Holds five slides
TRUSTED_SUSTAINABILITY
Specifications
Type Slide Mailer
Capacity (Metric) 5 L
Color White
Height (Metric) 18 mm
Length (Metric) 82 mm
Width (Metric) 20 mm