missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Slfn9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144357
This item is not returnable.
View return policy
Description
Slfn9 Polyclonal Antibody for Western Blot, ELISA
Endoribonuclease that cleaves tRNAs and rRNAs.
Specifications
| Slfn9 | |
| Polyclonal | |
| Unconjugated | |
| Slfn9 | |
| EC 3.1.-.-; Schlafen family member 9; Schlafen-9; Slfn9 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 237886 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 2.66 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| B1ARD6 | |
| Slfn9 | |
| Recombinant Mouse Schlafen family member 9 protein (1-83AA). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction