missing translation for 'onlineSavingsMsg'
Learn More

SLC4A1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16896005 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16896005 150 μL 150µL
16806015 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16896005

Brand: Proteintech 281311AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The Band 3 protein is the most abundant integral protein of human erythrocyte membranes and functions as the major anion transporter polypeptide of erythrocytes. Band 3 is a 90-100 kDa protein, its microheterogeneity is ascribed mainly to the structural heterogeneity of the carbohydrate moiety. The C-terminal portion of the molecule spans the membrane several times, and is responsible for anion transport. The N-terminal portion, comprising almost half of the polypeptide chain of Band 3, is located inside the red cell and interacts with cytoskeletal and cytoplasmic proteins such as ankyrin.

Specifications

Antigen SLC4A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SLC4A1
Gene Accession No. P02730, P04919
Gene Alias AE 1, AE1, Anion exchange protein 1, Anion exchanger 1, Band 3 anion transport protein, BND3, CD233, DI, EMPB3, EPB3, FR, RTA1A, SLC4A1, SW, WD, WD1, WR
Gene Symbols SLC4A1
Host Species Rabbit
Immunogen SLC4A1 Fusion Protein Ag27878
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20533, 6521
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.