missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SLC25A11 Recombinant Protein (P01)
Brand: Abnova™ H00008402-P01.25ug
Additional Details : Weight : 0.00010kg
Specifications
AAH16294 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
60.28 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MAATASAGAGGIDGKPRTSPKSVKFLFGGLAGMGATVFVQPLDLVKNRMQLSGEGAKTREYKTSFHALTSILKAEGLRGIYTGLSAGLLRQATYTTTRLGIYTVLFERLTGADGTPPGFLLKAVIGMTAGATGAFVGTPAEVALIRMTADGRLPADQRRGYKNVFNALIRITREEGVLTLWRGCIPTMARAVVVNAAQLASYSQSKQFLLDSGYFSDNILCHFCASMISGLVTTAASMPVDIAKTRIQNMRMIDGKPEYKNGLDVLFKVVRYEGFFSLWKGFTPYYARLGPHTVLTFIFLEQMNKAYKRLFLSG | |
OGC/SLC20A4 | |
SLC25A11 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
8402 | |
SLC25A11 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SLC25A11 | |
Human | |
Recombinant | |
Solution |