missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ SLC25A11 Recombinant Protein (P01)

Brand:  Abnova™ H00008402-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16132422

  • 4115.00 NOK / 10µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

AAH16294
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
60.28
8
Glutathione Sepharose 4 Fast Flow
10 ug
MAATASAGAGGIDGKPRTSPKSVKFLFGGLAGMGATVFVQPLDLVKNRMQLSGEGAKTREYKTSFHALTSILKAEGLRGIYTGLSAGLLRQATYTTTRLGIYTVLFERLTGADGTPPGFLLKAVIGMTAGATGAFVGTPAEVALIRMTADGRLPADQRRGYKNVFNALIRITREEGVLTLWRGCIPTMARAVVVNAAQLASYSQSKQFLLDSGYFSDNILCHFCASMISGLVTTAASMPVDIAKTRIQNMRMIDGKPEYKNGLDVLFKVVRYEGFFSLWKGFTPYYARLGPHTVLTFIFLEQMNKAYKRLFLSG
OGC/SLC20A4
SLC25A11
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
8402
SLC25A11 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SLC25A11
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers