Learn More
SLC22A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
Antigen SLC22A1
Concentration 0.27 mg/mL
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16803115
View Documents
Proteintech
24617-1-AP-20UL
20 μL
1710.33 NOK
20µL
View alternative products
Save Up to 
»
16893105
View Documents
Proteintech
24617-1-AP-150UL
150 μL
4836.10 NOK
150µL
View alternative products
Save Up to 
»
Description
Description
Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. Two transcript variants encoding two different isoforms have been found for this gene, but only the longer variant encodes a functional transporter.
Specifications
Specifications
SLC22A1
Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Mouse, Rat, Human
O08966, O15245, Q63089
20517, 24904, 6580
SLC22A1 Fusion Protein Ag18806
Primary
-20°C
SLC22A1
0.27 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
HOCT1, OCT1, oct1_cds, Organic cation transporter 1, SLC22A1
Slc22a1
IgG
Antigen Affinity Chromatography