missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC12A6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA580005
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
SLC12A6 is a member of the K-Cl cotransporter (KCC) family. K-Cl cotransporters are integral membrane proteins that lower intracellular chloride concentrations below the electrochemical equilibrium potential. The proteins encoded by this gene are activated by cell swelling induced by hypotonic conditions. Alternate splicing results in multiple transcript variants encoding different isoforms. Mutations in this gene are associated with agenesis of the corpus callosum with peripheral neuropathy.Specifications
SLC12A6 | |
Polyclonal | |
Unconjugated | |
SLC12A6 | |
9530023I19Rik; ACCPN; DKFZP434D2135; Electroneutral potassium-chloride cotransporter 3; furosemide-sensitive KCl cotransporter 3; gaxp; Kcc3; KCC3A; KCC3B; K-Cl cotransporter 3; potassium chloride cotransporter 3; potassium chloride cotransporter KCC3a-S3; potassium-chloride transporter-3a; potassium-chloride transporter-3b; SLC12A6; solute carrier family 12 (potassium/chloride transporter), member 6; solute carrier family 12 (potassium/chloride transporters), member 6; solute carrier family 12 member 6; solute carrier family 12, member 6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
107723, 691209, 9990 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5MG BSA and 0.05MG sodium azide, 0.05MG thimerosal | |
Q924N4, Q9UHW9 | |
SLC12A6 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human SLC12A6 (1060-1080aa QKAKSMEGFQDLLNMRPDQSN). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |